MilliporeSigma
  • Perovskite alkali metal samarium borohydrides: crystal structures and thermal decomposition.
Perovskite alkali metal samarium borohydrides: crystal structures and thermal decomposition.
Dalton transactions (Cambridge, England : 2003) (2017-08-30)
Kasper T Møller, Mathias Jørgensen, Alexander S Fogh, Torben R Jensen
ABSTRACT
A new synthesis method of samarium borohydride, Sm(BH