MilliporeSigma
  • Fast kinetics of magnesium monochloride cations in interlayer-expanded titanium disulfide for magnesium rechargeable batteries.
Fast kinetics of magnesium monochloride cations in interlayer-expanded titanium disulfide for magnesium rechargeable batteries.
Nature communications (2017-08-25)
Hyun Deog Yoo, Yanliang Liang, Hui Dong, Junhao Lin, Hua Wang, Yisheng Liu, Lu Ma, Tianpin Wu, Yifei Li, Qiang Ru, Yan Jing, Qinyou An, Wu Zhou, Jinghua Guo, Jun Lu, Sokrates T Pantelides, Xiaofeng Qian, Yan Yao
ABSTRACT
Magnesium rechargeable batteries potentially offer high-energy density, safety, and low cost due to the ability to employ divalent, dendrite-free, and earth-abundant magnesium metal anode. Despite recent progress, further development remains stagnated mainly due to the sluggish scission of magnesium-chloride bond and slow diffusion of divalent magnesium cations in cathodes. Here we report a battery chemistry that utilizes magnesium monochloride cations in expanded titanium disulfide. Combined theoretical modeling, spectroscopic analysis, and electrochemical study reveal fast diffusion kinetics of magnesium monochloride cations without scission of magnesium-chloride bond. The battery demonstrates the reversible intercalation of 1 and 1.7 magnesium monochloride cations per titanium at 25 and 60 °C, respectively, corresponding to up to 400 mAh g
Sigma-Aldrich
1-Butyl-1-methylpyrrolidinium chloride, ≥99% (T)