MilliporeSigma
  • Activity of carboxylesterases, glutathione-S-transferase and monooxygenase on Rhipicephalus microplus exposed to fluazuron.
Activity of carboxylesterases, glutathione-S-transferase and monooxygenase on Rhipicephalus microplus exposed to fluazuron.
Parasitology international (2017-04-25)
Fabrício Nascimento Gaudêncio, Guilherme Marcondes Klafke, Vinícius Menezes Tunholi-Alves, Thaís Paes Ferreira, Cristiane Nunes Coelho, Adivaldo Henrique da Fonseca, Isabele da Costa Angelo, Jairo Pinheiro
ABSTRACT
The objective of this study was to assess the effect of the exposure to fluazuron on the activity of common pesticide detoxification enzyme groups in the cattle tick (Rhipicephalus microplus). Engorged females of a susceptible strain (POA) and a resistant strain (Jaguar) were exposed in vitro to fluazuron and their eggs and larvae were used to compare the activities of the general esterases, mixed-function oxidases (MFO) and glutathione-S-transferase (GST). The results showed significant elevation in MFO contents and esterases activity in the resistant strain when compared with the susceptible strain, in eggs and larvae respectively. In the POA strain, the MFO activity in eggs was down-regulated by fluazuron exposure. Based on these results, it can be concluded that different detoxification enzymes can act in distinct pathways depending on the tick's development stage, and may be related to fluazuron detoxification in resistant strains.
Product Description
Supelco
Fluazuron, PESTANAL®, analytical standard
Pricing and availability is not currently available.