MilliporeSigma
  • pH-Induced Switchable Superwettability of Efficient Antibacterial Fabrics for Durable Selective Oil/Water Separation.
pH-Induced Switchable Superwettability of Efficient Antibacterial Fabrics for Durable Selective Oil/Water Separation.
ACS applied materials & interfaces (2017-08-15)
Yuchen Fu, Biyu Jin, Qinghua Zhang, Xiaoli Zhan, Fengqiu Chen
ABSTRACT
The superhydrophobic antibacterial fabrics with intelligent switchable wettability were fabricated by the cross-link reaction among pH-responsive antibacterial copolymer tethered hydroxyl groups, methylol-contained poly(ureaformaldehyde) nanoparticles (PUF NPs), and hexamethylene diisocyanate. It was found that the surface concentration of N
Sigma-Aldrich
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, contains inhibitor, 97%
Pricing and availability is not currently available.