MilliporeSigma
  • Temperature-responsive nanogel multilayers of poly(N-vinylcaprolactam) for topical drug delivery.
Temperature-responsive nanogel multilayers of poly(N-vinylcaprolactam) for topical drug delivery.
Journal of colloid and interface science (2017-08-02)
Oleksandra Zavgorodnya, Carlos A Carmona-Moran, Veronika Kozlovskaya, Fei Liu, Timothy M Wick, Eugenia Kharlampieva
ABSTRACT
We report nanothin temperature-responsive hydrogel films of poly(N-vinylcaprolactam) nanoparticles (νPVCL) with remarkably high loading capacity for topical drug delivery. Highly swollen (νPVCL)
Sigma-Aldrich
N-Vinylcaprolactam, 98%, stabilized