MilliporeSigma
  • Collecting near mature and immature orchid seeds for ex situ conservation: 'in vitro collecting' as a case study.
Collecting near mature and immature orchid seeds for ex situ conservation: 'in vitro collecting' as a case study.
Botanical studies (2017-08-10)
Jonathan P Kendon, Landy Rajaovelona, Helen Sandford, Rui Fang, Jake Bell, Viswambharan Sarasan
ABSTRACT
Lack of phenological information and efficient collecting methods are considered impediments for orchid seed collecting. This leads to opportunistic collecting as part of general seed collecting schedules that may last few weeks especially in remote areas. The study explored the feasibility of collecting near mature and immature seeds to support conservation action plans. Mature, near mature and immature seeds of orchids were collected from the wild in the Central Highlands of Madagascar (CHM). Seed capsules were collected in sterile culture medium in the wild, to prevent deterioration of seeds inside the capsule after collecting, later to be cultured under laboratory conditions. Seed capsules collected by the in vitro collecting (IVC) method were kept in very good condition for up to 4 weeks before germination under in vitro conditions. Significantly faster and higher germination rate (p < 0.001) than mature seeds was observed in a number of taxa collected during a 3 year-long study. In some taxa even immature seeds, with no apparent sign of testa covering the embryo, germinated following IVC where mature seeds failed to germinate. We propose that IVC method has potential to complement conventional seed collecting by increasing the germplasm that can be used in integrated conservation action plans. Improvements can be made in developing collections for taxa from biodiversity hotspots and remote areas where collecting requires considerable resources. This method can further be used on a wider selection of plants from different geographic areas and on embryo rescue programmes for economically important plants.
Sigma-Aldrich
Sodium dichloroisocyanurate, 96%