MilliporeSigma
  • Expression of β-globin by cancer cells promotes cell survival during blood-borne dissemination.
Expression of β-globin by cancer cells promotes cell survival during blood-borne dissemination.
Nature communications (2017-02-10)
Yu Zheng, David T Miyamoto, Ben S Wittner, James P Sullivan, Nicola Aceto, Nicole Vincent Jordan, Min Yu, Nezihi Murat Karabacak, Valentine Comaills, Robert Morris, Rushil Desai, Niyati Desai, Erin Emmons, John D Milner, Richard J Lee, Chin-Lee Wu, Lecia V Sequist, Wilhelm Haas, David T Ting, Mehmet Toner, Sridhar Ramaswamy, Shyamala Maheswaran, Daniel A Haber
ABSTRACT
Metastasis-competent circulating tumour cells (CTCs) experience oxidative stress in the bloodstream, but their survival mechanisms are not well defined. Here, comparing single-cell RNA-Seq profiles of CTCs from breast, prostate and lung cancers, we observe consistent induction of β-globin (HBB), but not its partner α-globin (HBA). The tumour-specific origin of HBB is confirmed by sequence polymorphisms within human xenograft-derived CTCs in mouse models. Increased intracellular reactive oxygen species (ROS) in cultured breast CTCs triggers HBB induction, mediated through the transcriptional regulator KLF4. Depletion of HBB in CTC-derived cultures has minimal effects on primary tumour growth, but it greatly increases apoptosis following ROS exposure, and dramatically reduces CTC-derived lung metastases. These effects are reversed by the anti-oxidant N-Acetyl Cysteine. Conversely, overexpression of HBB is sufficient to suppress intracellular ROS within CTCs. Altogether, these observations suggest that β-globin is selectively deregulated in cancer cells, mediating a cytoprotective effect during blood-borne metastasis.
Sigma-Aldrich
Poly-Prep Slides, poly-L-lysine coated glass slides
Pricing and availability is not currently available.