MilliporeSigma
  • Highly Ordered Mesostructured Vanadium Phosphonate toward Electrode Materials for Lithium-Ion Batteries.
Highly Ordered Mesostructured Vanadium Phosphonate toward Electrode Materials for Lithium-Ion Batteries.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-03-08)
Peng Mei, Malay Pramanik, Jaewoo Lee, Yusuke Ide, Zeid Abdullah Alothman, Jung Ho Kim, Yusuke Yamauchi
ABSTRACT
Highly ordered mesostructured vanadium phosphonates (VP) have been synthesized in the presence of cetyltrimethylammonium bromide (CTAB) as a structure-directing agent. Nitrilotris(methylene)triphosphonic acid (NMPA) and (ammonium/sodium) metavanadate (NH
Sigma-Aldrich
N-(3-Methoxypropyl)acrylamide, contains MEHQ as inhibitor, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrilotris(methylene)triphosphonic acid solution, 50 wt. % in H2O
Pricing and availability is not currently available.