MilliporeSigma
  • Deubiquitinating enzyme USP22 positively regulates c-Myc stability and tumorigenic activity in mammalian and breast cancer cells.
Deubiquitinating enzyme USP22 positively regulates c-Myc stability and tumorigenic activity in mammalian and breast cancer cells.
Journal of cellular physiology (2017-02-06)
Dongyeon Kim, Ahyoung Hong, Hye In Park, Woo Hyun Shin, Lang Yoo, Seo Jeong Jeon, Kwang Chul Chung
ABSTRACT
The proto-oncogene c-Myc has a pivotal function in growth control, differentiation, and apoptosis and is frequently affected in human cancer, including breast cancer. Ubiquitin-specific protease 22 (USP22), a member of the USP family of deubiquitinating enzymes (DUBs), mediates deubiquitination of target proteins, including histone H2B and H2A, telomeric repeat binding factor 1, and cyclin B1. USP22 is also a component of the mammalian SAGA transcriptional co-activating complex. In this study, we explored the functional role of USP22 in modulating c-Myc stability and its physiological relevance in breast cancer progression. We found that USP22 promotes deubiquitination of c-Myc in several breast cancer cell lines, resulting in increased levels of c-Myc. Consistent with this, USP22 knockdown reduces c-Myc levels. Furthermore, overexpression of USP22 stimulates breast cancer cell growth and colony formation, and increases c-Myc tumorigenic activity. In conclusion, the present study reveals that USP22 in breast cancer cell lines increases c-Myc stability through c-Myc deubiquitination, which is closely correlated with breast cancer progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human USP22