MilliporeSigma
  • Development of a UV crosslinked biodegradable hydrogel containing adipose derived stem cells to promote vascularization for skin wounds and tissue engineering.
Development of a UV crosslinked biodegradable hydrogel containing adipose derived stem cells to promote vascularization for skin wounds and tissue engineering.
Biomaterials (2017-03-28)
Gozde Eke, Naside Mangir, Nesrin Hasirci, Sheila MacNeil, Vasif Hasirci
ABSTRACT
The aim of this study was to design a dermal substitute containing adipose derived stem cells (ADSC) that can be used to improve the regeneration of skin on difficult wound beds by stimulating rapid neovascularization. This was achieved by first synthesizing methacrylated gelatin (GelMA) and methacrylated hyaluronic acid (HAMA) precursors which could be stored at -80
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%