MilliporeSigma
  • Chloride channel ClC- 2 enhances intestinal epithelial tight junction barrier function via regulation of caveolin-1 and caveolar trafficking of occludin.
Chloride channel ClC- 2 enhances intestinal epithelial tight junction barrier function via regulation of caveolin-1 and caveolar trafficking of occludin.
Experimental cell research (2017-02-06)
Prashant K Nighot, Lana Leung, Thomas Y Ma
ABSTRACT
Previous studies have demonstrated that the chloride channel ClC-2 plays a critical role in intestinal epithelial tight junction (TJ) barrier function via intracellular trafficking of TJ protein occludin. To study the mechanism of ClC-2-mediated TJ barrier function and intracellular trafficking of occludin, we established ClC-2 over-expressing Caco-2 cell line (Caco-2
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture