MilliporeSigma
  • Endogenous hydrogen sulfide contributes to uterine quiescence during pregnancy.
Endogenous hydrogen sulfide contributes to uterine quiescence during pregnancy.
Reproduction (Cambridge, England) (2017-02-12)
Xingji You, Zixi Chen, Huina Zhao, Chen Xu, Weina Liu, Qianqian Sun, Ping He, Hang Gu, Xin Ni
ABSTRACT
Recent evidence suggests that uterine activation for labor is associated with inflammation within uterine tissues. Hydrogen sulfide (H
Sigma-Aldrich
MISSION® esiRNA, targeting human CBS
Pricing and availability is not currently available.