MilliporeSigma
  • Co-gasification of bituminous coal and hydrochar derived from municipal solid waste: Reactivity and synergy.
Co-gasification of bituminous coal and hydrochar derived from municipal solid waste: Reactivity and synergy.
Bioresource technology (2017-05-26)
Juntao Wei, Qinghua Guo, Qing He, Lu Ding, Kunio Yoshikawa, Guangsuo Yu
ABSTRACT
In this work, the influences of gasification temperature and blended ratio on co-gasification reactivity and synergy of Shenfu bituminous coal (SF) and municipal solid waste-derived hydrochar (HTC) were investigated using TGA. Additionally, active alkaline and alkaline earth metal (AAEM) transformation during co-gasification was quantitatively analyzed by inductively coupled plasma optical emission spectrometer for correlating synergy on co-gasification reactivity. The results showed that higher char gasification reactivity existed at higher HTC char proportion and gasification temperature, and the main synergy behaviour on co-gasification reactivity was performed as synergistic effect. Enhanced synergistic effect at lower temperature was mainly resulted from more obviously inhibiting the primary AAEM (i.e. active Ca) transformation, and weak synergistic effect still existed at higher temperature since more active K with prominent catalysis was retained. Furthermore, more active HTC-derived AAEM remaining in SF sample during co-gasification would lead to enhanced synergistic effect as HTC char proportion increased.
Sigma-Aldrich
2-(Methacryloyloxy)ethyl acetoacetate, 95%, contains BHT as stabilizer
Pricing and availability is not currently available.