MilliporeSigma
  • Determination of pyrrolizidine alkaloids in selected feed materials with gas chromatography-mass spectrometry.
Determination of pyrrolizidine alkaloids in selected feed materials with gas chromatography-mass spectrometry.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2017-03-10)
Ewelina Kowalczyk, Krzysztof Kwiatek
ABSTRACT
1,2-Dehydropyrrolizidine alkaloids are known to be toxic to many animals and humans. To provide safety of feeds a method based on gas chromatography-mass spectrometry enabling the determination of a content of 1,2-unsaturated PAs in feed materials was developed. After extraction with aqueous solution of HCl and purification of the extract, 1,2-unsaturated alkaloids are reduced to their common backbone structures and subsequently derivatised with heptafluorobutyric anhydride (HFBA). The method was validated according to SANTE/11945/2015. All received parameters are consistent with the document requirements as recovery of a final compound retronecine derivative was from 81.8% to 94.4% when retrosine was used for spiking and from 72.7% to 85.5% when retrorsine N-oxide was spiked. The repeatability was calculated as relative standards deviation and ranged from 7.5% to 14.4%, for N-oxide was from 7.9% to 15.4%. The reproducibility was in the range from 14.2% to 16.3% and from 17.0% to 18.1% for free base and N-oxide respectively. The limit of quantification was determined as 10 µg kg