MilliporeSigma
  • Probing spermiogenesis: a digital strategy for mouse acrosome classification.
Probing spermiogenesis: a digital strategy for mouse acrosome classification.
Scientific reports (2017-06-18)
Alessandro Taloni, Francesc Font-Clos, Luca Guidetti, Simone Milan, Miriam Ascagni, Chiara Vasco, Maria Enrica Pasini, Maria Rosa Gioria, Emilio Ciusani, Stefano Zapperi, Caterina A M La Porta
ABSTRACT
Classification of morphological features in biological samples is usually performed by a trained eye but the increasing amount of available digital images calls for semi-automatic classification techniques. Here we explore this possibility in the context of acrosome morphological analysis during spermiogenesis. Our method combines feature extraction from three dimensional reconstruction of confocal images with principal component analysis and machine learning. The method could be particularly useful in cases where the amount of data does not allow for a direct inspection by trained eye.
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture