MilliporeSigma
  • A multicenter, placebo-controlled, double-blind clinical trial assessing the effects of a multicomponent nutritional supplement for treating photoaged skin in healthy women.
A multicenter, placebo-controlled, double-blind clinical trial assessing the effects of a multicomponent nutritional supplement for treating photoaged skin in healthy women.
Journal of cosmetic dermatology (2016-10-27)
Jay E Birnbaum, David H McDaniel, Janet Hickman, Lisa Dispensa, Anne Le Moigne, Larry Buchner
ABSTRACT
Dietary or supplementary intake of nutrients and other ingredients positively affects skin appearance. Evaluate a multicomponent nutritional supplement on photoaged skin. This multicenter, double-blind, placebo-controlled trial randomized healthy 35- to 65-year-old women with Fitzpatrick skin types I-IV and Glogau classification types II-III to a multicomponent nutritional supplement or placebo for 24 weeks. The primary endpoint was Investigator Global Assessment of overall facial appearance at week 24. Secondary endpoints included investigator- and subject-rated assessments of the face, décolletage, and hands; facial photography assessments (conducted by an independent panel of dermatologists); and instrumental measures. Analysis of variance was used to assess between-group differences (P ≤ 0.05). Of 194 randomized subjects, 171 completed the study. Subjects had a mean age of 53 years and were primarily white (81%), had Glogau II (58%) and Fitzpatrick III (45%; significantly more supplement subjects had Fitzpatrick III [54%] vs. placebo [35%]; P = 0.039). At week 24, Investigator Global Assessment of overall facial appearance was numerically but not statistically better for supplement over placebo (mean difference: 0.14 [95% confidence interval: -0.16-0.44]; P = 0.358). A significant treatment-by-site interaction (P = 0.073) was observed; by-site analyses revealed a significant difference at one of three geographical sites for supplement vs. placebo (P = 0.001). Differences on secondary endpoints were generally not significant. In the first multicenter study conducted with this multicomponent nutritional supplement, no significantly greater effects vs. placebo were observed on the primary endpoint, perhaps because of significant between-site variability.