MilliporeSigma
  • Kruppel-like factor 4 inhibits non-small cell lung cancer cell growth and aggressiveness by stimulating transforming growth factor-β1-meidated ERK/JNK/NF-κB signaling pathways.
Kruppel-like factor 4 inhibits non-small cell lung cancer cell growth and aggressiveness by stimulating transforming growth factor-β1-meidated ERK/JNK/NF-κB signaling pathways.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2017-06-21)
Renzhi Yu, Lei Han, Xin Ni, Minghuan Wang, Ping Xue, Li Zhang, Mei Yuan
ABSTRACT
Non-small cell lung cancer is one of the most common epithelial tumors that cause the most common cancer-related mortality due to invasive ability. Research has found that Kruppel-like factor 4, a zinc-finger transcription factor, plays a critical role in the tumor evolution and progression. However, the molecular signal pathways mediated by Kruppel-like factor 4 in the progression of non-small cell lung cancer cells have not been well understood yet. In this study, we investigated the possible role and potential mechanism of Kruppel-like factor 4 in growth and aggressiveness of non-small cell lung cancer cells. Results showed that Kruppel-like factor 4 is downregulated in non-small cell lung cancer cells. Here, we found that Kruppel-like factor 4 knockdown promoted growth and aggressiveness of non-small cell lung cancer cells, as well as enhanced apoptotic resistance induced by tunicamycin. We also found that Kruppel-like factor 4 overexpression significantly suppressed growth and aggressiveness of non-small cell lung cancer cells. Apoptosis rate of non-small cell lung cancer cells induced by tunicamycin was promoted by Kruppel-like factor 4 overexpression. Kruppel-like factor 4 overexpression inhibited transforming growth factor-β1, extracellular signal-regulated protein kinase, C-jun N-terminal kinase, and nuclear factor-κB expression levels in non-small cell lung cancer cells. Mechanistically, Kruppel-like factor 4-mediated tumorigenesis involved suppression of a transforming growth factor-β1-meidated extracellular signal-regulated protein kinase/C-jun N-terminal kinase/nuclear factor-κB transcriptional program in non-small cell lung cancer cells. Our results revealed that Kruppel-like factor 4 overexpression non-small cell lung cancer cell reduces tumor growth in experimental mice. Overall, these data indicate the inhibitory role of Kruppel-like factor 4 in non-small cell lung cancer cells and elaborate a potential molecular signal pathway involving in growth and aggressiveness. Findings identify Kruppel-like factor 4 can be regarded as a possible new molecular agent for designing novel therapeutic protein drug for lung cancer treatment to control non-small cell lung cancer growth.
Sigma-Aldrich
MISSION® esiRNA, targeting human KLF4