MilliporeSigma
  • Short-term regular aerobic exercise reduces oxidative stress produced by acute in the adipose microvasculature.
Short-term regular aerobic exercise reduces oxidative stress produced by acute in the adipose microvasculature.
American journal of physiology. Heart and circulatory physiology (2017-02-27)
Austin T Robinson, Ibra S Fancher, Varadarajan Sudhahar, Jing Tan Bian, Marc D Cook, Abeer M Mahmoud, Mohamed M Ali, Masuko Ushio-Fukai, Michael D Brown, Tohru Fukai, Shane A Phillips
ABSTRACT
High blood pressure has been shown to elicit impaired dilation in the vasculature. The purpose of this investigation was to elucidate the mechanisms through which high pressure may elicit vascular dysfunction and determine the mechanisms through which regular aerobic exercise protects arteries against high pressure. Male C57BL/6J mice were subjected to 2 wk of voluntary running (~6 km/day) for comparison with sedentary controls. Hindlimb adipose resistance arteries were dissected from mice for measurements of flow-induced dilation (FID; with or without high intraluminal pressure exposure) or protein expression of NADPH oxidase II (NOX II) and superoxide dismutase (SOD). Microvascular endothelial cells were subjected to high physiological laminar shear stress (20 dyn/cm
Sigma-Aldrich
VAS2870, ≥97% (HPLC)