MilliporeSigma
  • Visual experience regulates the development of long-term synaptic modifications induced by low-frequency stimulation in mouse visual cortex.
Visual experience regulates the development of long-term synaptic modifications induced by low-frequency stimulation in mouse visual cortex.
Neuroscience research (2017-03-13)
Taketoshi Sugimura, Mariko Yamamoto, Kazumasa Yamada, Yukio Komatsu, Yumiko Yoshimura
ABSTRACT
Manipulation of visual experience can considerably modify visual responses of visual cortical neurons even in adulthood in the mouse, although the modification is less profound than that observed during the critical period. Our previous studies demonstrated that low-frequency (2Hz) stimulation for 15min applied to layer 4 induces T-type Ca
Product Description
Supelco
Micro particle size standard based on polystyrene monodisperse, analytical standard, size: 30.0 μm