MilliporeSigma
  • Direct and convenient measurement of plasmid stability in lab and clinical isolates of E. coli.
Direct and convenient measurement of plasmid stability in lab and clinical isolates of E. coli.
Scientific reports (2017-07-08)
Siyi Chen, Mårten Larsson, Robert C Robinson, Swaine L Chen
ABSTRACT
Plasmids are important mobile elements in bacteria, contributing to evolution, virulence, and antibiotic resistance. Natural plasmids are generally large and maintained at low copy number and thus prone to be lost. Therefore, dedicated plasmid maintenance systems have evolved, leading to plasmid loss rates as low as 1 per 10
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG