MilliporeSigma
Singlet oxygen-responsive micelles for enhanced photodynamic therapy.
Journal of controlled release : official journal of the Controlled Release Society (2017-05-22)
Xiaodan Li, Min Gao, Keting Xin, Ling Zhang, Dan Ding, Deling Kong, Zheng Wang, Yang Shi, Fabian Kiessling, Twan Lammers, Jianjun Cheng, Yanjun Zhao
ABSTRACT
Photodynamic therapy (PDT) efficacy is limited by the very short half-life and limited diffusion radius of singlet oxygen (
Sigma-Aldrich
1-(3-Aminopropyl)imidazole, ≥97%