MilliporeSigma
Downregulation of RBO-PI4KIIIα Facilitates Aβ
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-04-21)
Xiao Zhang, Wen-An Wang, Li-Xiang Jiang, Hai-Yan Liu, Bao-Zhu Zhang, Nastasia Lim, Qing-Yi Li, Fu-De Huang
ABSTRACT
Phosphoinositides and their metabolizing enzymes are involved in Aβ
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine, without calcium chloride and sodium bicarbonate, powder, suitable for insect cell culture
Pricing and availability is not currently available.