MilliporeSigma
Histamine regulates murine primary dendritic cell functions.
Immunopharmacology and immunotoxicology (2016-08-26)
Heiko Schenk, Detlef Neumann, Christina Kloth
ABSTRACT
The modulation of antigen uptake and activation of dendritic cells (DCs) by histamine may function as a regulator of inflammation. Therefore, we sought to determine the impact of histamine on antigen uptake by and activation of murine DCs. DCs from spleen and lung were either identified by flow cytometry or were immunomagnetically enriched. Cells were stimulated with histamine, and the regulation of MHC-II and co-stimulatory molecule expression (CD80, CD86, and ICOS-L) and antigen uptake were quantified by flow cytometry. Individual contributions of the histamine receptor subtypes were determined by using the antagonists mepyramine (histamine H1-receptor: H1R), famotidine (H2R), and JNJ 7777120 (H4R). Histamine accelerated the uptake of soluble antigen via the H1R, H2R, and H4R in splenic DCs. Co-stimulatory molecule expression was enhanced already by enrichment procedures, thus, the analyses were performed in unseparated cell populations. Histamine enhanced the expression of CD86 and ICOS-L while expression of CD80 was unaffected. Antagonism at H1R, H2R, and H4R and at H1R and H4R reduced the histamine-induced enhanced expression of CD86 and ICOS-L, respectively. Histamine contributes to the regulation of the immunological synapse by stimulation of antigen uptake and activation of DCs via H1R, H2R, and H4R.
Sigma-Aldrich
JNJ7777120, ≥98% (HPLC)
Pricing and availability is not currently available.