MilliporeSigma
  • Sodium hypochlorite as an alternative to hydrogen peroxide in Fenton process for industrial scale.
Sodium hypochlorite as an alternative to hydrogen peroxide in Fenton process for industrial scale.
Water research (2017-05-20)
Jamshid Behin, Abbas Akbari, Mohsen Mahmoudi, Mehdi Khajeh
ABSTRACT
In present work, the treatment of aromatic compounds of simulated wastewater was performed by Fenton and NaOCl/Fe
Sigma-Aldrich
2,4-Diaminotoluene, 98%