MilliporeSigma
  • An LC-MS/MS method for the simultaneous determination of 15 antipsychotics and two metabolites in hair and its application to rat hair.
An LC-MS/MS method for the simultaneous determination of 15 antipsychotics and two metabolites in hair and its application to rat hair.
Forensic science international (2017-01-24)
Juhyun Sim, Eunmi Kim, Wonkyung Yang, Sanghee Woo, Sangwhan In
ABSTRACT
In recent years, the inappropriate use of antipsychotics by young Korean men has become a social problem. As military service exemptions are given for mental illness, some men pose as mental health patients to avoid military service. In order to verify the authenticity of mental illnesses, we developed simultaneous analytical methods for the detection of 15 antipsychotics and 2 of their metabolites in hair using liquid chromatography-tandem mass spectrometry (LC-MS/MS) analysis. The target drugs were modafinil, atomoxetine, aripiprazole, benztropine, buspirone, duloxetine, gabapentin, oxcarbazepine, topiramate, escitalopram, paliperidone, ziprasidone, lamotrigine, clonazepam, levetiracetam, and metabolites of oxcarbazepine and clonazepam. To remove possible contaminants on the hair surface, hair samples were washed twice with methanol and distilled water, and then were extracted with methanol overnight at 38°C. Desipramine-d
Product Description
Supelco
10,11-Dihydro-10-hydroxycarbamazepine, analytical standard