MilliporeSigma
  • Diversity and Activity of Diazotrophs in Great Barrier Reef Surface Waters.
Diversity and Activity of Diazotrophs in Great Barrier Reef Surface Waters.
Frontiers in microbiology (2017-06-24)
Lauren F Messer, Mark V Brown, Miles J Furnas, Richard L Carney, A D McKinnon, Justin R Seymour
ABSTRACT
Discrepancies between bioavailable nitrogen (N) concentrations and phytoplankton growth rates in the oligotrophic waters of the Great Barrier Reef (GBR) suggest that undetermined N sources must play a significant role in supporting primary productivity. One such source could be biological dinitrogen (N
Sigma-Aldrich
Nitrogen-14N2, 99.99 atom % 14N