MilliporeSigma
Ganoderic Acid A Metabolites and Their Metabolic Kinetics.
Frontiers in pharmacology (2017-03-23)
Fang-Rui Cao, Li Feng, Lin-Hu Ye, Li-Sha Wang, Bing-Xin Xiao, Xue Tao, Qi Chang
ABSTRACT
Ganoderic acid A (GAA), a representative active triterpenoid from
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%