MilliporeSigma
  • New enantioselective LC method development and validation for the assay of modafinil.
New enantioselective LC method development and validation for the assay of modafinil.
Journal of pharmaceutical and biomedical analysis (2017-02-24)
Marianna Harvanová, Taťána Gondová
ABSTRACT
For the first time, a new, fast and sensitive chromatographic method for the separation and determination of modafinil enantiomers was developed on chiral stationary phase with macrocyclic glycopeptide teicoplanin in the polar organic mode. The effect of the mobile phase composition and column temperature on the retention and enantioseparation were studied. The separation was performed using a Chirobiotic T column (250×4.6mm, 5μm) with methanol and triethylamine (100/0.05, v/v) as the mobile phase at a flow rate of 1.0mL/min, and UV detection at 225nm. The total analysis time is less than 6min, which is faster than the previous chiral HPLC methods (total run time of 12min). Calibration curves were linear (R
Sigma-Aldrich
Armodafinil, ≥98% (HPLC)
Pricing and availability is not currently available.