MilliporeSigma
  • Transparent conductive electrodes from graphene/PEDOT:PSS hybrid inks for ultrathin organic photodetectors.
Transparent conductive electrodes from graphene/PEDOT:PSS hybrid inks for ultrathin organic photodetectors.
Advanced materials (Deerfield Beach, Fla.) (2014-12-03)
Zhaoyang Liu, Khaled Parvez, Rongjin Li, Renhao Dong, Xinliang Feng, Klaus Müllen
ABSTRACT
A novel solution fabrication of large-area, highly conductive graphene films by spray-coating of a hybrid ink of exfoliated graphene (EG)/poly(3,4-ethylenedioxythiophene):poly(styrenesulfonate) (PEDOT:PSS) (PH1000) is demonstrated. The fabricated graphene films exhibit excellent mechanical properties, thus enabling their application as bottom electrodes in ultrathin organic photodetector devices with performance comparable to that of the state-of-the-art Si-based inorganic photodetectors.
Sigma-Aldrich
Graphene/PEDOT:PSS hybrid ink, dispersion in DMF, avg. no. of layers, 1 ‑ 3