MilliporeSigma
  • Structural and magnetic properties of semiquinonate based Al(iii) and Ga(iii) complexes.
Structural and magnetic properties of semiquinonate based Al(iii) and Ga(iii) complexes.
Dalton transactions (Cambridge, England : 2003) (2017-01-11)
Chinmoy Das, Pragya Shukla, Lorenzo Sorace, Maheswaran Shanmugam
ABSTRACT
The reaction of anhydrous MCl
Sigma-Aldrich
3,5-Di-tert-butyl-o-benzoquinone, 98%
Pricing and availability is not currently available.