MilliporeSigma
  • (p-Amidinophenyl)methanesulfonyl fluoride, an irreversible inhibitor of serine proteases.
(p-Amidinophenyl)methanesulfonyl fluoride, an irreversible inhibitor of serine proteases.
Biochemistry (1980-10-14)
R Laura, D J Robison, D H Bing
ABSTRACT
p-(Amidinophenyl)methanesulfonyl fluoride (p-APMSF) has been synthesized and shown to be a specific, irreversible inhibitor of the class of plasma serine proteases which demonstrate substrate specificity for the positively charged side chains of the amino acid lysine or arginine. In equimolar concentration, this compound causes immediate and complete irreversible inhibition of bovine trypsin and human thrombin. A 5-10-fold molar excess of reagent over enzyme is required to achieve complete irreversible inhibition of bovine Factor Xa, human plasmin, human C1-r, and human C1-s. the Ki of p-APMSF for all of the above-mentioned proteases is between 1 and 2 microM. In contrast, p-APMSF in large molar excess does not inactivate chymotrypsin or acetylcholinesterase. The unique reactivity of p-APMSF has been further shown in comparison with the related compound p-nitrophenyl (p-amidinophenyl)methanesulfonate which is an active-site titrant for thrombin but reacts poorly with Factor Xa, C1-r, and C1-s and is not hydrolyzed by bovine trypsin or human plasmin. Similarly, (p-amidinophenyl)methanesulfonate has a Ki of 30 microM for thrombin but is a poor inhibitor of trypsin, Factor Xa, C1-r, C1-s, and plasmin. Studies with bovine trypsin have demonstrated that the inhibitory activity of p-APMSF is the result of its interaction with the diisopropyl fluorophosphate reactive site. The unique reactivity of this inhibitor classifies it as one of the most effective active site directed reagents for this class of serine proteases. Collectively, these results suggest that the primary substrate binding site of these enzymes, which share a high degree of structural homology, do in fact significantly differ from each other in their ability to interact with low molecular weight inhibitors and synthetic substrates.