MilliporeSigma
  • Activation of G protein-coupled receptor 30 by thiodiphenol promotes proliferation of estrogen receptor α-positive breast cancer cells.
Activation of G protein-coupled receptor 30 by thiodiphenol promotes proliferation of estrogen receptor α-positive breast cancer cells.
Chemosphere (2016-11-24)
Bingli Lei, Wei Peng, Gang Xu, Minghong Wu, Yu Wen, Jie Xu, Zhiqiang Yu, Yipei Wang
ABSTRACT
Many studies have been shown that environmental estrogen bisphenol A (BPA) can activate nuclear receptor (estrogen receptor alpha, ERα) or membrane receptor (G-protein-coupled receptor, GPR30) in breast cancer cells and exerts genomic or nongenomic actions inducing cell proliferation. 4,4'-thiodiphenol (TDP) as one of BPA derivatives exhibits more potent estrogenic activity than BPA does. However, comparatively little is known about the ways in which TDP interferes with these signaling pathways and produces cell biological changes. This study evaluated the effect of TDP on cell viability, reactive oxygen species (ROS) formation, and intercellular calcium (Ca
Sigma-Aldrich
4,4′-Thiodiphenol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Pricing and availability is not currently available.