MilliporeSigma
  • Foldable interpenetrated metal-organic frameworks/carbon nanotubes thin film for lithium-sulfur batteries.
Foldable interpenetrated metal-organic frameworks/carbon nanotubes thin film for lithium-sulfur batteries.
Nature communications (2017-03-07)
Yiyin Mao, Gaoran Li, Yi Guo, Zhoupeng Li, Chengdu Liang, Xinsheng Peng, Zhan Lin
ABSTRACT
Lithium-sulfur batteries are promising technologies for powering flexible devices due to their high energy density, low cost and environmental friendliness, when the insulating nature, shuttle effect and volume expansion of sulfur electrodes are well addressed. Here, we report a strategy of using foldable interpenetrated metal-organic frameworks/carbon nanotubes thin film for binder-free advanced lithium-sulfur batteries through a facile confinement conversion. The carbon nanotubes interpenetrate through the metal-organic frameworks crystal and interweave the electrode into a stratified structure to provide both conductivity and structural integrity, while the highly porous metal-organic frameworks endow the electrode with strong sulfur confinement to achieve good cyclability. These hierarchical porous interpenetrated three-dimensional conductive networks with well confined S