MilliporeSigma
  • Substituted Phthalimide AC94377 Is a Selective Agonist of the Gibberellin Receptor GID1.
Substituted Phthalimide AC94377 Is a Selective Agonist of the Gibberellin Receptor GID1.
Plant physiology (2016-12-03)
Kai Jiang, Masato Otani, Hiroaki Shimotakahara, Jung-Min Yoon, Seung-Hyun Park, Tomoko Miyaji, Takeshi Nakano, Hidemitsu Nakamura, Masatoshi Nakajima, Tadao Asami
ABSTRACT
Gibberellin (GA) is a major plant hormone that regulates plant growth and development and is widely used as a plant growth regulator in agricultural production. There is an increasing demand for function-limited GA mimics due to the limitations on the agronomical application of GA to crops, including GA's high cost of producing and its leading to the crops' lodging. AC94377, a substituted phthalimide, is a chemical that mimics the growth-regulating activity of GAs in various plants, despite its structural difference. Although AC94377 is widely studied in many weeds and crops, its mode of action as a GA mimic is largely unknown. In this study, we confirmed that AC94377 displays GA-like activities in Arabidopsis (Arabidopsis thaliana) and demonstrated that AC94377 binds to the Arabidopsis GIBBERELLIN INSENSITIVE DWARF1 (GID1) receptor (AtGID1), forms the AtGID1-AC94377-DELLA complex, and induces the degradation of DELLA protein. Our results also indicated that AC94377 is selective for a specific subtype among three AtGID1s and that the selectivity of AC94377 is attributable to a single residue at the entrance to the hydrophobic pocket of GID1. We conclude that AC94377 is a GID1 agonist with selectivity for a specific subtype of GID1, which could be further developed and used as a function-limited regulator of plant growth in both basic study and agriculture.
Product Description
Supelco
Prohexadione-Calcium, PESTANAL®, analytical standard
Pricing and availability is not currently available.