MilliporeSigma
  • Occurrence, fate and environmental risk of linear alkylbenzene sulfonate in the Langat and Selangor River basins, Malaysia.
Occurrence, fate and environmental risk of linear alkylbenzene sulfonate in the Langat and Selangor River basins, Malaysia.
Chemosphere (2017-01-13)
Nobumitsu Sakai, Junichi Shirasaka, Yasuto Matsui, Mohd Redzuan Ramli, Kousuke Yoshida, Mustafa Ali Mohd, Minoru Yoneda
ABSTRACT
Five homologs (C
Sigma-Aldrich
4-Octylbenzenesulfonic acid sodium salt, 97%
Pricing and availability is not currently available.