MilliporeSigma
  • A simple and fast Double-Flow microfluidic device based liquid-phase microextraction (DF-µLPME) for the determination of parabens in water samples.
A simple and fast Double-Flow microfluidic device based liquid-phase microextraction (DF-µLPME) for the determination of parabens in water samples.
Talanta (2017-02-06)
María Ramos-Payan, Santiago Maspoch, Andreu Llobera
ABSTRACT
A fast double-flow microfluidic based liquid phase microextraction (DF-µLPME) combined with a HPLC-UV procedure using diode array detection has been developed for the determination of the four most widely used parabens: Ethyl 4-hydroxybenzoate (EtP), Propyl 4-hydroxybenzoate (PrP), Butyl 4-hydroxybenzoate (BuP) and IsoButyl 4-hydroxybenzoate (iBuP) in water samples. Parabens have successfully been determined in environmental (lake and river water) samples with excellent clean up, high extraction efficiency and good enrichment factor using double-flow conditions. The microfluidic device consists of two micro-channels, which contain the acceptor and sample solution separated by a flat membrane (support liquid membrane). The sample (0.32mM HCl) and acceptor phase (5.6mM NaOH) are delivered to the µLPME at 10µLmin
Sigma-Aldrich
Dihexyl ether, 97%