MilliporeSigma
  • Mitofusin-2 determines mitochondrial network architecture and mitochondrial metabolism. A novel regulatory mechanism altered in obesity.
Mitofusin-2 determines mitochondrial network architecture and mitochondrial metabolism. A novel regulatory mechanism altered in obesity.
The Journal of biological chemistry (2003-02-25)
Daniel Bach, Sara Pich, Francesc X Soriano, Nathalie Vega, Bernhard Baumgartner, Josep Oriola, Jens R Daugaard, Jorge Lloberas, Marta Camps, Juleen R Zierath, Rémi Rabasa-Lhoret, Harriet Wallberg-Henriksson, Martine Laville, Manuel Palacín, Hubert Vidal, Francisca Rivera, Martin Brand, Antonio Zorzano
ABSTRACT
In many cells and specially in muscle, mitochondria form elongated filaments or a branched reticulum. We show that Mfn2 (mitofusin 2), a mitochondrial membrane protein that participates in mitochondrial fusion in mammalian cells, is induced during myogenesis and contributes to the maintenance and operation of the mitochondrial network. Repression of Mfn2 caused morphological and functional fragmentation of the mitochondrial network into independent clusters. Concomitantly, repression of Mfn2 reduced glucose oxidation, mitochondrial membrane potential, cell respiration, and mitochondrial proton leak. We also show that the Mfn2-dependent mechanism of mitochondrial control is disturbed in obesity by reduced Mfn2 expression. In all, our data indicate that Mfn2 expression is crucial in mitochondrial metabolism through the maintenance of the mitochondrial network architecture, and reduced Mfn2 expression may explain some of the metabolic alterations associated with obesity.