MilliporeSigma
  • Iridium/monodentate phosphoramidite catalyzed asymmetric hydrogenation of N-aryl imines.
Iridium/monodentate phosphoramidite catalyzed asymmetric hydrogenation of N-aryl imines.
Journal of the American Chemical Society (2009-06-18)
Natasa Mrsić, Adriaan J Minnaard, Ben L Feringa, Johannes G de Vries
ABSTRACT
Asymmetric hydrogenation of N-aryl acetophenone imines using iridium/PipPhos leads to very high enantioselectivities up to >99% depending on the presence of electron-donating substituents in the 2-, 3-, and 5-position of the aryl ring. If the substituent is 2-methoxy, the resultant secondary amines are easily oxidatively deprotected using trichloroisocyanuric acid to give the primary amines in good yield with full retention of enantioselectivity.