MilliporeSigma
  • Monomeric Cinchona Alkaloid-Based Catalysts for Highly Enantioselective Bromolactonisation of Alkynes.
Monomeric Cinchona Alkaloid-Based Catalysts for Highly Enantioselective Bromolactonisation of Alkynes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-10-12)
Michael Wilking, Constantin G Daniliuc, Ulrich Hennecke
ABSTRACT
The cinchona alkaloid dimer (DHQD)2 PHAL has been shown to be a broadly applicable catalyst for asymmetric halogenations. However, this catalyst does not have to be dimeric and a class of monomeric quinidine and quinine-derived catalysts was prepared, often showing superior selectivity in bromolactonisations of terminal alkynoic acids. Mechanistic investigations show that these organocatalysts act as host molecules that can bind carboxylic acid-based substrates as guests with substantial binding constants. Based on these findings, it is proposed that this class of catalysts is bifunctional in nature activating the halogenating agent as well as the nucleophile in electrophilic halogenation reactions.