MilliporeSigma
  • Keratin mediates the recognition of apoptotic and necrotic cells through dendritic cell receptor DEC205/CD205.
Keratin mediates the recognition of apoptotic and necrotic cells through dendritic cell receptor DEC205/CD205.
Proceedings of the National Academy of Sciences of the United States of America (2016-11-09)
Longxing Cao, Haishuang Chang, Xiangyi Shi, Chao Peng, Yongning He
ABSTRACT
Clearance of dead cells is critical for maintaining homeostasis and prevents autoimmunity and inflammation. When cells undergo apoptosis and necrosis, specific markers are exposed and recognized by the receptors on phagocytes. DEC205 (CD205) is an endocytotic receptor on dendritic cells with antigen presentation function and has been widely used in immune therapies for vaccine generation. It has been shown that human DEC205 recognizes apoptotic and necrotic cells in a pH-dependent fashion. However, the natural ligand(s) of DEC205 remains unknown. Here we find that keratins are the cellular ligands of human DEC205. DEC205 binds to keratins specifically at acidic, but not basic, pH through its N-terminal domains. Keratins form intermediate filaments and are important for maintaining the strength of cells and tissues. Our results suggest that keratins also function as cell markers of apoptotic and necrotic cells and mediate a pH-dependent pathway for the immune recognition of dead cells.
Sigma-Aldrich
Dicyanocobinamide, ≥93%