MilliporeSigma
  • A novel electrochemical biomimetic sensor based on poly(Cu-AMT) with reduced graphene oxide for ultrasensitive detection of dopamine.
A novel electrochemical biomimetic sensor based on poly(Cu-AMT) with reduced graphene oxide for ultrasensitive detection of dopamine.
Talanta (2016-11-14)
Yaru Li, Yue Gu, Bo Zheng, Lan Luo, Cong Li, Xiaoyi Yan, Tingting Zhang, Nannan Lu, Zhiquan Zhang
ABSTRACT
A polymerized film of copper-2-amino-5-mercapto-1,3,4-thiadiazole (Cu(II)-AMT) complex (poly(Cu-AMT)) was successfully achieved via a simple and low-cost electrochemical methodology. Subsequently, a noncovalent nanohybrid of poly(Cu-AMT) with reduced graphene oxide (rGO) (rGO-poly(Cu-AMT)) was prepared through π-π stacking interaction as an efficient mimetic enzyme for the ultrasensitive and selective detection of dopamine (DA). The rGO-poly(Cu-AMT) nanocomposites showed considerable mimetic enzyme catalytic activity, which may be attributed to the significant promotion of the electron transfer between the substrate and graphene-based carbon materials, and also the synergistic electrocatalytic effect in mimetic enzyme between rGO sheet and poly(Cu-AMT). The electrocatalytic and sensing performances of the biomimetic sensor based on the rGO-poly(Cu-AMT) nanocomposites were evaluated in detail. The biomimetic sensor enables a reliable and sensitive determination of DA with a linear range of 0.01-40μM and a detection limit of 3.48nM at a signal-to-noise ratio of 3. In addition, we applied the proposed method to detect DA in real sample with satisfactory results. Accordingly, the rGO-poly(Cu-AMT) is one of the promising mimetic enzyme for electrocatalysis and biosensing.