MilliporeSigma
  • Determination of the concentrations of alternative antifouling agents on the Korean coast.
Determination of the concentrations of alternative antifouling agents on the Korean coast.
Marine pollution bulletin (2016-10-04)
Seongeon Lee, Yong-Woo Lee
ABSTRACT
Since the ban on tributyltin (TBT) in 2008, dozens of alternative antifouling agents have been used instead. Unfortunately, these alternative antifouling agents contain diverse toxic components, which have contaminated the coasts. From 2006 to 2013, the concentration of chlorothalonil, dichlofluanid, and Irgarol 1051 were monitored continuously, and their mean concentrations increased due to the increase in maritime cargo and consequent increase in the incoming and outgoing ships in harbors. An analysis of the sampling points according to harbor type showed that the contamination was more severe in fishing and big harbors, where there are more incoming and outgoing ships. A correlation analysis indicated a high correlation between chlorothalonil and dichlofluanid. Unlike Irgarol 1051, which is only used as an antifouling agent, the other two substances are used as agricultural chemicals as well, which could explain the high correlation. This study suggests that these alternative antifouling agents should be tracked continuously.
Product Description
Supelco
Vinclozolin, PESTANAL®, analytical standard
Pricing and availability is not currently available.