MilliporeSigma
  • Clinical value of eukaryotic elongation factor 2 (eEF2) in non-small cell lung cancer patients.
Clinical value of eukaryotic elongation factor 2 (eEF2) in non-small cell lung cancer patients.
Asian Pacific journal of cancer prevention : APJCP (2014-01-01)
Hong-Gang Sun, Xue-Jun Dong, Tao Lu, Ming-Feng Yang, Xing-Mu Wang
ABSTRACT
The purpose of this study was to evaluate a new type of tumor biomarker, eukaryotic elongation factor 2 (eEF2), in serum for the early diagnosis, confirmative diagnosis as well as assessment of treatment of non-small cell lung cancer (NSCLC). 130 patients with NSCLC and 50 healthy individuals undergoing physical examination in our hospital provided the observation and healthy control groups. An enzyme linked immune sorbent assay (ELISA) method was applied to determine serum eEF2 levels. Serum neuron specific enolase (NSE) and squamous cell carcinoma antigen (SCC) levels in the observation group were assessed with an automatic biochemical analyzer. The median levels of eEF2 in the serum of NSCLC patients was found to be significantly higher than the healthy control group (p < 0.01) and it was markedly higher in stages III, IV than stages I, II (p < 0.05). eEF2 was higher with tumor size ≥ 2 cm than <2 cm (P< 0.01). Furthermore, two weeks after surgery patients showed a significant trend for eEF2 decrease (p < 0.05). The eukaryotic elongation factor 2 (eEF2) has certain clinical values for early diagnosis, verification, and prognosis as well as classification of lung cancer patients.