MilliporeSigma
  • Healthy and pre-eclamptic placental basal plate lining cells: quantitative comparisons based on confocal laser scanning microscopy.
Healthy and pre-eclamptic placental basal plate lining cells: quantitative comparisons based on confocal laser scanning microscopy.
Microscopy research and technique (2004-08-03)
R K Smith, C D Ockleford, S Byrne, P Bosio, R Sanders
ABSTRACT
Immunocytochemical confocal laser scanning microscope images of the monolayer of cells lining the intervillus space at the basal plate of term placentae were analysed using stereology. Immunoreactively-distinct regions of this mosaic layer were measured. In basal plate from healthy pregnancies, trophoblast epithelium occupied 18.91% of the surface area and endothelium 60.81%. In pre-eclampsia the equivalent areas were 15.57% and 67.63%. Acellular fibrinoid covers the remaining area and this component decreases in area in pre-eclampsia. The statistically significant increase in the cellular endothelial compartment may be relevant to the hypertensive pathology of pre-eclampsia as endothelial signalling plays a major role in regulation of blood pressure.