MilliporeSigma
  • Selective targeting of the α5-subunit of GABAA receptors relaxes airway smooth muscle and inhibits cellular calcium handling.
Selective targeting of the α5-subunit of GABAA receptors relaxes airway smooth muscle and inhibits cellular calcium handling.
American journal of physiology. Lung cellular and molecular physiology (2015-02-11)
George Gallos, Gene T Yocum, Matthew E Siviski, Peter D Yim, Xiao Wen Fu, Michael M Poe, James M Cook, Neil Harrison, Jose Perez-Zoghbi, Charles W Emala
ABSTRACT
The clinical need for novel bronchodilators for the treatment of bronchoconstrictive diseases remains a major medical issue. Modulation of airway smooth muscle (ASM) chloride via GABAA receptor activation to achieve relaxation of precontracted ASM represents a potentially beneficial therapeutic option. Since human ASM GABAA receptors express only the α4- and α5-subunits, there is an opportunity to selectively target ASM GABAA receptors to improve drug efficacy and minimize side effects. Recently, a novel compound (R)-ethyl8-ethynyl-6-(2-fluorophenyl)-4-methyl-4H-benzo[f]imidazo[1,5-a][1,4] diazepine-3-carboxylate (SH-053-2'F-R-CH3) with allosteric selectivity for α5-subunit containing GABAA receptors has become available. We questioned whether this novel GABAA α5-selective ligand relaxes ASM and affects intracellular calcium concentration ([Ca(2+)]i) regulation. Immunohistochemical staining localized the GABAA α5-subunit to human ASM. The selective GABAA α5 ligand SH-053-2'F-R-CH3 relaxes precontracted intact ASM; increases GABA-activated chloride currents in human ASM cells in voltage-clamp electrophysiology studies; and attenuates bradykinin-induced increases in [Ca(2+)]i, store-operated Ca(2+) entry, and methacholine-induced Ca(2+) oscillations in peripheral murine lung slices. In conclusion, selective subunit targeting of endogenous α5-subunit containing GABAA receptors on ASM may represent a novel therapeutic option to treat severe bronchospasm.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GABA A Receptor α 5 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.