MilliporeSigma
  • PARP6 acts as a tumor suppressor via downregulating Survivin expression in colorectal cancer.
PARP6 acts as a tumor suppressor via downregulating Survivin expression in colorectal cancer.
Oncotarget (2016-03-05)
Guangying Qi, Yasusei Kudo, Bo Tang, Tian Liu, Shengjian Jin, Jing Liu, Xiaoxu Zuo, Sisi Mi, Wenhuan Shao, Xiaojuan Ma, Takaaki Tsunematsu, Naozumi Ishimaru, Sien Zeng, Masaaki Tatsuka, Fumio Shimamoto
ABSTRACT
Poly (ADP-ribose) polymerases (PARPs) are enzymes that transfer ADP-ribose groups to target proteins and are involved in a variety of biological processes. PARP6 is a novel member, and our previous findings suggest that PARP6 may act as a tumor suppressor via suppressing cell cycle progression. However, it is still unclear that PARP6 function besides growth suppression in colorectal cancer (CRC). In this study, we examined tumor suppressive roles of PAPR6 in CRC cells both in vitro and in vivo. We found that PARP6 inhibited colony formation, invasion and migration as well as cell proliferation. Moreover, ectopic overexpression of PARP6 decreased Survivin expression, which acts as an oncogene and is involved in apoptosis and mitosis. We confirmed the inverse correlation between PARP6 and Survivin expression in CRC cases by immunohistochemistry. Importantly, CRC cases with downregulation of PARP6 and upregulation of Survivin showed poor prognosis. In summary, PARP6 acts as a tumor suppressor via downregulating Survivin expression in CRC. PARP6 can be a novel diagnostic and therapeutic target together with Survivin for CRC.
Sigma-Aldrich
Anti-PARP6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PARP6 Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)