MilliporeSigma
  • The BRCA1/BARD1-interacting protein OLA1 functions in centrosome regulation.
The BRCA1/BARD1-interacting protein OLA1 functions in centrosome regulation.
Molecular cell (2013-12-03)
Ayako Matsuzawa, Shin-Ichiro Kanno, Masahiro Nakayama, Hironori Mochiduki, Leizhen Wei, Tatsuro Shimaoka, Yumiko Furukawa, Kei Kato, Shun Shibata, Akira Yasui, Chikashi Ishioka, Natsuko Chiba
ABSTRACT
The breast and ovarian cancer-specific tumor suppressor BRCA1, along with its heterodimer partner BRCA1-associated RING domain protein (BARD1), plays important roles in DNA repair, centrosome regulation, and transcription. To explore further functions of BRCA1/BARD1, we performed mass spectrometry analysis and identified Obg-like ATPase 1 (OLA1) as a protein that interacts with the carboxy-terminal region of BARD1. OLA1 directly bound to the amino-terminal region of BRCA1 and γ-tubulin. OLA1 localized to centrosomes in interphase and to the spindle pole in mitotic phase, and its knockdown resulted in centrosome amplification and the activation of microtubule aster formation. OLA1 with a mutation observed in breast cancer cell line, E168Q, failed to bind BRCA1 and rescue the OLA1 knockdown-induced centrosome amplification. BRCA1 variant I42V also abrogated the binding of BRCA1 to OLA1. These findings suggest that OLA1 plays an important role in centrosome regulation together with BRCA1.
Sigma-Aldrich
Anti-OLA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.