MilliporeSigma
  • Age-associated declines in mitochondrial biogenesis and protein quality control factors are minimized by exercise training.
Age-associated declines in mitochondrial biogenesis and protein quality control factors are minimized by exercise training.
American journal of physiology. Regulatory, integrative and comparative physiology (2012-05-11)
Erika Koltai, Nikolett Hart, Albert W Taylor, Sataro Goto, Jenny K Ngo, Kelvin J A Davies, Zsolt Radak
ABSTRACT
A decline in mitochondrial biogenesis and mitochondrial protein quality control in skeletal muscle is a common finding in aging, but exercise training has been suggested as a possible cure. In this report, we tested the hypothesis that moderate-intensity exercise training could prevent the age-associated deterioration in mitochondrial biogenesis in the gastrocnemius muscle of Wistar rats. Exercise training, consisting of treadmill running at 60% of the initial Vo(2max), reversed or attenuated significant age-associated (detrimental) declines in mitochondrial mass (succinate dehydrogenase, citrate synthase, cytochrome-c oxidase-4, mtDNA), SIRT1 activity, AMPK, pAMPK, and peroxisome proliferator-activated receptor gamma coactivator 1-α, UCP3, and the Lon protease. Exercise training also decreased the gap between young and old animals in other measured parameters, including nuclear respiratory factor 1, mitochondrial transcription factor A, fission-1, mitofusin-1, and polynucleotide phosphorylase levels. We conclude that exercise training can help minimize detrimental skeletal muscle aging deficits by improving mitochondrial protein quality control and biogenesis.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-UCP-3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution