MilliporeSigma
  • Deregulation of ARID1A, CDH1, cMET and PIK3CA and target-related microRNA expression in gastric cancer.
Deregulation of ARID1A, CDH1, cMET and PIK3CA and target-related microRNA expression in gastric cancer.
Oncotarget (2015-09-04)
Maider Ibarrola-Villava, Marta J Llorca-Cardeñosa, Noelia Tarazona, Cristina Mongort, Tania Fleitas, José Alejandro Perez-Fidalgo, Susana Roselló, Samuel Navarro, Gloria Ribas, Andrés Cervantes
ABSTRACT
Genetic and epigenetic alterations play an important role in gastric cancer (GC) pathogenesis. Aberrations of the phosphatidylinositol-3-kinase signaling pathway are well described. However, emerging genes have been described such as, the chromatin remodeling gene ARID1A. Our aim was to determine the expression levels of four GC-related genes, ARID1A, CDH1, cMET and PIK3CA, and 14 target-related microRNAs (miRNAs). We compared mRNA and miRNA expression levels among 66 gastric tumor and normal adjacent mucosa samples using quantitative real-time reverse transcription PCR. Moreover, ARID1A, cMET and PIK3CA protein levels were assessed by immunohistochemistry (IHC). Finally, gene and miRNAs associations with clinical characteristics and outcome were also evaluated. An increased cMET and PIK3CA mRNA expression was found in 78.0% (P = 2.20 × 10-5) and 73.8% (P = 1.00 × 10-3) of the tumors, respectively. Moreover, IHC revealed that cMET and PIK3CA expression was positive in 63.6% and 87.8% of the tumors, respectively. Six miRNAs had significantly different expression between paired-samples, finding five up-regulated [miR-223-3p (P = 1.65 × 10-6), miR-19a-3p (P = 1.23 × 10-4), miR-128-3p (P = 3.49 × 10-4), miR-130b-3p (P = 1.00 × 10-3) and miR-34a-5p (P = 4.00 × 10-3)] and one down-regulated [miR-124-3p (P = 0.03)]. Our data suggest that cMET, PIK3CA and target-related miRNAs play an important role in GC and may serve as potential targets for therapy.
Sigma-Aldrich
Anti-ARID1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution