MilliporeSigma
  • α2δ-1 signaling in nucleus accumbens is necessary for cocaine-induced relapse.
α2δ-1 signaling in nucleus accumbens is necessary for cocaine-induced relapse.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-06-21)
Sade Spencer, Robyn M Brown, Gabriel C Quintero, Yonatan M Kupchik, Charles A Thomas, Kathryn J Reissner, Peter W Kalivas
ABSTRACT
Relapse to cocaine seeking is associated with potentiated excitatory synapses in nucleus accumbens. α2δ-1 is an auxiliary subunit of voltage-gated calcium channels that affects calcium-channel trafficking and kinetics, initiates extracellular signaling cascades, and promotes excitatory synaptogenesis. Previous data demonstrate that repeated exposure to alcohol, nicotine, methamphetamine, and morphine upregulates α2δ-1 in reward-related brain regions, but it was unclear whether this alteration generalized to cocaine. Here, we show that α2δ-1 protein was increased in nucleus accumbens after cocaine self-administration and extinction compared with saline controls. Furthermore, the endogenous ligand thrombospondin-1, responsible for the synaptogenic properties of the α2δ-1 receptor, was likewise elevated. Using whole-cell patch-clamp recordings of EPSCs in nucleus accumbens, we demonstrated that gabapentin, a specific α2δ-1 antagonist, preferentially reduced the amplitude and increased the paired-pulse ratio of EPSCs evoked by electrical stimulation in slices from cocaine-experienced rats compared with controls. In vivo, gabapentin microinjected in the nucleus accumbens core attenuated cocaine-primed but not cue-induced reinstatement. Importantly, gabapentin's effects on drug seeking were not due to a general depression of spontaneous or cocaine-induced locomotor activity. Moreover, gabapentin had no effect on reinstatement of sucrose seeking. These data indicate that α2δ-1 contributes specifically to cocaine-reinstated drug seeking, and identifies this protein as a target for the development of cocaine relapse medications. These results also inform ongoing discussion in the literature regarding efficacy of gabapentin as a candidate addiction therapy.
Sigma-Aldrich
Monoclonal Anti-Dihydropyridine Receptor (α2 Subunit) antibody produced in mouse, clone 20A, ascites fluid, buffered aqueous solution
Pricing and availability is not currently available.